Skip to Content
Merck
All Photos(1)

Key Documents

Y0002007

Sulfobutylbetadex impurity B

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1,4-Butane sultone, 4-Hydroxybutane-1-sulfonic acid δ-sultone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O3S
CAS Number:
Molecular Weight:
136.17
Beilstein:
110588
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

betadex

manufacturer/tradename

EDQM

refractive index

n20/D 1.464 (lit.)

bp

>165 °C/25 mmHg (lit.)

mp

12-15 °C (lit.)

density

1.331 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

O=S1(=O)CCCCO1

InChI

1S/C4H8O3S/c5-8(6)4-2-1-3-7-8/h1-4H2

InChI key

MHYFEEDKONKGEB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Sulfobutylbetadex impurity B EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

309.2 °F

Flash Point(C)

154 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service