Skip to Content
Merck
All Photos(1)

Key Documents

NIST909C

Human Serum

NIST® SRM® 909c

Synonym(s):

Blood Serum from Humans

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

packaging

pkg of 3 x 2 mL

manufacturer/tradename

NIST®

technique(s)

mass spectrometry (MS): suitable

application(s)

clinical testing

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

A unit of NIST SRM 909C human serum consists of three vials, each containing approximately 2 mL of frozen human serum.

SRM 909C_Cert

SRM 909C_SDS

Application

The human blood serum standard reference material (SRM) is meant primarily for use in validating analytical methods for the determination of specified constituents in human serum. This SRM can also be used for quality assurance when assigning values to in-house control materials.

Features and Benefits

  • The certified values are traceable to the International System of Units (SI) for the amount of substance and mass, expressed both in mass concentrations (milligrams per deciliter, micrograms per deciliter, nanograms per milliliter, or grams per liter) and amount concentration (nanomoles per liter or micromoles per liter)

Other Notes

  • Refer to the certificate for information on expiry, safety, usage, and storage.
  • Notes on biomaterials, disposal, and transport information are available in SDS.
Certified for the analytes listed below.
Cholesterol, Creatinine, Glycerides, Urea, Calcium (Ca), Chloride (Cl), Magnesium (Mg), Potassium (K), Sodium (Na), Iron (Fe), Selenium (Se), Transferrin (Tf)

Reference/Informational value is provided for the following analytes.
Total Protein, Disialo Tf, Trisialo Tf, Tetrasialo Tf, Pentasialo Tf, Hexasialo Tf

See certificate for values and more details at nist.gov/SRM.

Analysis Note

Endotoxin tested

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-sensitivity programmable flow method for assessment of total antioxidant capacity in biological samples
Sara S. M, et al.
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 261-266 (2016)
Mahmoud Elhusseiny Mostafa et al.
Journal of chromatography. A, 1670, 462952-462952 (2022-03-27)
LC-MS metabolomic analysis in complex biological matrices may be complicated by degeneracy when using large-bore columns. Degeneracy is the detection of multiple mass spectral peaks from the same analyte due to adduction of salts to the metabolite, dimerization, or loss
Sara S Marques et al.
International journal of molecular sciences, 15(7), 11387-11402 (2014-06-27)
Total antioxidant capacity assays are recognized as instrumental to establish antioxidant status of biological samples, however the varying experimental conditions result in conclusions that may not be transposable to other settings. After selection of the complexing agent, reagent addition order

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service