Skip to Content
Merck
All Photos(1)

Key Documents

F0180000

Fludrocortisone acetate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

9α-Fluoro-11β,17α,21-trihydroxy-4-pregnene-3,20-dione acetate, 9-Fluorocortisol acetate, 9-Fluorohydrocortisone acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H31FO6
CAS Number:
Molecular Weight:
422.49
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

fludrocortisone

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC(=O)OCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C

InChI

1S/C23H31FO6/c1-13(25)30-12-19(28)22(29)9-7-16-17-5-4-14-10-15(26)6-8-20(14,2)23(17,24)18(27)11-21(16,22)3/h10,16-18,27,29H,4-9,11-12H2,1-3H3/t16-,17-,18-,20-,21-,22-,23-/m0/s1

InChI key

SYWHXTATXSMDSB-GSLJADNHSA-N

Gene Information

human ... NR3C2(4306)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Fludrocortisone acetate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Fludrocortisone acetate is a synthetic corticosteroid with more mineralocorticoid than glucocorticoid activity.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hitoshi Ishiguro et al.
Cancer chemotherapy and pharmacology, 74(2), 249-255 (2014-06-02)
A synthetic glucocorticoid, dexamethasone, was recently shown to inhibit bladder cancer cell invasion and metastasis through the glucocorticoid receptor (GR) pathway but increased cell proliferation via inhibiting apoptosis particularly induced by cisplatin. Therefore, comedication with dexamethasone in bladder cancer patients
C Nagelreiter et al.
International journal of pharmaceutics, 478(2), 429-438 (2014-12-06)
In order to customise treatment for patients, topical formulations are often diluted with drug-free cream bases to adjust the drug dose and thereby the formulations' activity to the patients' needs. However, the process of dilution influences properties of the formulations.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service