Skip to Content
Merck

BP080

Chloroxylenol

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

4-Chloro-3,5-dimethylphenol, 4-Chloro-3,5-xylenol, 4-Chloro-sym-m-xylenol, PCMX

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
ClC6H2(CH3)2OH
CAS Number:
Molecular Weight:
156.61
EC Number:
201-793-8
UNSPSC Code:
41116107
NACRES:
NA.24
Beilstein/REAXYS Number:
1862539
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

chloroxylenol

form

crystalline powder

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

mp

114-116 °C (lit.)

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

Cc1cc(O)cc(C)c1Cl

InChI

1S/C8H9ClO/c1-5-3-7(10)4-6(2)8(5)9/h3-4,10H,1-2H3

InChI key

OSDLLIBGSJNGJE-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Chloroxylenol BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Also used in monographs such as:
  • Chloroxylenol Solution
  • Packaging

    Unit quantity: 500 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

    Other Notes

    Sales restrictions may apply.


    Still not finding the right product?

    Explore all of our products under Chloroxylenol


    pictograms

    Exclamation mark

    signalword

    Warning

    Hazard Classifications

    Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

    Storage Class

    11 - Combustible Solids

    wgk

    WGK 1



    Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

    If you need assistance, please contact Customer Support

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library