Skip to Content
Merck
All Photos(1)

Key Documents

ARK2091

SAFC

Clomipramine hydrochloride

BP, EP, USP

Pharma Manufacturing

Synonym(s):

3-Chloro-10,11-dihydro-N,N-dimethyl-5H-dibenz[b,f]azepine-5-propanamine hydrochloride, Anafranil hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H23ClN2 · HCl
CAS Number:
Molecular Weight:
351.31
MDL number:
UNSPSC Code:
51292917

Agency

BP
EP
USP

Quality Level

storage temp.

room temp

SMILES string

Cl.CN(C)CCCN1c2ccccc2CCc3ccc(Cl)cc13

InChI

1S/C19H23ClN2.ClH/c1-21(2)12-5-13-22-18-7-4-3-6-15(18)8-9-16-10-11-17(20)14-19(16)22;/h3-4,6-7,10-11,14H,5,8-9,12-13H2,1-2H3;1H

InChI key

WIMWMKZEIBHDTH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT SE 1 - STOT SE 3

Target Organs

Central nervous system, Central nervous system,Cardiovascular

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service