Skip to Content
Merck
All Photos(2)

Key Documents

49376-U

Sigma-Aldrich

ProClin 150

ProClin preservative for use in IVD manufacturing, CMIT/MIT 1.5%, pkg of 50 mL

Synonym(s):

1.5% solution of 5-chloro-2-methyl-4-isothiazolin-3-one and 2-Methyl-4-isothiazolin-3-one

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352201
NACRES:
NA.21

Quality Level

description

biocide for IVD use

form

liquid

shelf life

2 yr

contains

23 to 25% Mg salts

composition

CMIT/MIT, 1.5%

packaging

pkg of 50 mL

parameter

operating temperature at <45 °C

matrix

Water

operating pH range

2.5-8.5

pH range

1.7-3.7

bp

100 °C

mp

-21 °C

solubility

water: soluble

density

1.20 g/cm3

Looking for similar products? Visit Product Comparison Guide

Application

ProClin preservatives are immediately microstatic upon contact with microbial organisms as a result of their ability to quickly penetrate cell membranes and inhibit specific enzymes crucial to cellular respiration. Multiple specific sites of ProClin toxicity prevent microorganisms from developing high levels of resistance, making ProClin highly efficient even over extended storage periods of your IVD components.

Legal Information

ProClin is a trademark of LANXESS Corp.

Disclaimer

The product is not intended for use as a biocide under global biocide regulations, including but not limited to US EPA′s Federal Insecticide Fungicide and Rodenticide Act, European Biocidal Products Regulation, Canada′s Pest Management Regulatory Agency, Turkey′s Biocidal Products Regulation, Korea′s Consumer Chemical Products and Biocide Safety Management Act (K-BPR) and others.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1C - Skin Sens. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Key raw materials for lateral flow assays optimize IVD assay development for successful commercialization.

Key raw materials for lateral flow assays optimize IVD assay development for successful commercialization.

Key raw materials for lateral flow assays optimize IVD assay development for successful commercialization.

Key raw materials for lateral flow assays optimize IVD assay development for successful commercialization.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service