Skip to Content
Merck
All Photos(2)

Key Documents

32451

Supelco

2-Hydroxyibuprofen

VETRANAL®, analytical standard

Synonym(s):

2-[4-(2-Hydroxy-2-methylpropyl)phenyl]propionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H18O3
CAS Number:
Molecular Weight:
222.28
Beilstein:
3051852
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

CC(C(O)=O)c1ccc(CC(C)(C)O)cc1

InChI

1S/C13H18O3/c1-9(12(14)15)11-6-4-10(5-7-11)8-13(2,3)16/h4-7,9,16H,8H2,1-3H3,(H,14,15)

InChI key

UJHKVYPPCJBOSG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Hydroxyibuprofen is a well-known human metabolite of ibuprofen. It finds applications as a nonsteroidal and anti-inflammatory drug.

Application

2-Hydroxyibuprofen may be used as a reference standard in the determination of 2-hydroxyibuprofen in urine samples using high-performance liquid chromatography (HPLC) and ultrasound-assisted emulsification microextraction (USAEME) with ultra-high-performance liquid chromatography-tandem mass spectrometry (UHPLC−MS/MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ultrasound-assisted emulsification microextraction combined with ultra-high performance liquid chromatography--tandem mass spectrometry for the analysis of ibuprofen and its metabolites in human urine.
Magiera S and Gulmez S
Journal of Pharmaceutical and Biomedical Analysis, 92(2), 193-202 (2014)
Simultaneous quantitative determination of the major phase I and II metabolites of ibuprofen in biological fluids by high-performance liquid chromatography on dynamically modified silica.
Kepp RD, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 696(2), 235-241 (1997)
Wastewater and Public Health: Bacterial and Pharmaceutical Exposures (2015)
Emilia Waraksa et al.
Journal of separation science, 41(20), 3881-3891 (2018-08-14)
Ibuprofen is widely used in human and veterinary medicine for the treatment of chronic pain as well as rheumatic and musculoskeletal disorders. However, the analgesic and anti-inflammatory properties of Ibuprofen have contributed to frequent drug abuse in equestrian sports. A

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service