Skip to Content
Merck
All Photos(2)

Key Documents

319112

Sigma-Aldrich

Methylene Blue solution

0.05 wt. % in H2O

Synonym(s):

Ehrlich’s reagent III, Loeffler’s Methylene Blue

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H18ClN3S
CAS Number:
Molecular Weight:
319.85
Colour Index Number:
52015
Beilstein:
3641570
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

liquid

Quality Level

concentration

0.05 wt. % in H2O

density

0.98 g/mL at 25 °C

λmax

665 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Cl-].CN(C)c1ccc2N=C3C=C\C(C=C3Sc2c1)=[N+](/C)C

InChI

1S/C16H18N3S.ClH/c1-18(2)11-5-7-13-15(9-11)20-16-10-12(19(3)4)6-8-14(16)17-13;/h5-10H,1-4H3;1H/q+1;/p-1

InChI key

CXKWCBBOMKCUKX-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Methylene Blue solution has been used to study structural alterations and p53 immune-expression in rats.

Biochem/physiol Actions

Methylene blue (MB) is a highly recognized histological dye. It plays a vital role in microbiology and pharmacology. It has been employed in staining living organisms and in methemoglobinemia treatment. MB exhibits maximum absorption at 664nm. Exposure to methylene blue leads to various health hazards such as increased heart rate, vomiting, shock, Heinz body formation, cyanosis, jaundice, quadriplegia and tissue necrosis in humans

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexandru Grigoroiu et al.
Scientific reports, 10(1), 3947-3947 (2020-03-05)
Hyperspectral imaging (HSI) is being explored in endoscopy as a tool to extract biochemical information that may improve contrast for early cancer detection in the gastrointestinal tract. Motion artefacts during medical endoscopy have traditionally limited HSI application, however, recent developments
Microscopic structural alterations
and p53 immune-expression in adult
rat testis in response to intra-tunical versus
intra-parenchymal injection of methylene blue
Mazroa S A
International Journal of Clinical and Experimental Pathology, 9, 5346-5356 (2016)
Methylene blue in photodynamic therapy: From basic mechanisms to clinical applications.
Tardivo JP
Photodiagnosis and Photodynamic Therapy, 2, 175-191 (2005)
J How et al.
Gynecologic oncology, 137(3), 436-442 (2015-04-15)
With the debate over extent of lymphadenectomy in endometrial cancer, sentinel lymph node (SLN) mapping may provide a focused approach to evaluate the most relevant lymph nodes (LN) while minimizing the complications. We evaluated SLN mapping using filtered technetium(99), indocyanine
Bram Vivijs et al.
Applied and environmental microbiology, 80(19), 6054-6061 (2014-07-27)
Some members of the family Enterobacteriaceae ferment sugars via the mixed-acid fermentation pathway. This yields large amounts of acids, causing strong and sometimes even lethal acidification of the environment. Other family members employ the 2,3-butanediol fermentation pathway, which generates comparatively

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service