Skip to Content
Merck
All Photos(1)

Key Documents

28713

Millipore

LB Broth

NutriSelect® Plus, from non-animal source(Vegitone), powder, pkg of 500 g, non-sterile, nonselective for Escherichia coli

Synonym(s):

E. coli enrichment broth, LB Broth, Vegitone, Luria Bertani Broth, Vegitone, Vegitone LB Broth

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171607
NACRES:
NA.85

biological source

non-animal source (Vegitone)

Quality Level

sterility

non-sterile

form

powder

shelf life

limited shelf life, expiry date on the label

composition

sodium chloride, 5.0 g/L
tryptone (vegetable), 10.0 g/L
yeast extract, 5.0 g/L

packaging

pkg of 500 g

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

color

clear to slightly opaque yellow to amber Prepared media
light yellow Dehydrated powder

final pH

7.0±0.2 (25 °C)

application(s)

agriculture
food and beverages
life science and biopharma
microbiology
pharmaceutical

suitability

nonselective for Escherichia coli (Recombinant)
nonselective for coliforms

Looking for similar products? Visit Product Comparison Guide

General description

Luria Bertani Broth Vegitone is free of animal derived material. It is used for the cultivation and maintenance of recombinant strains of Escherichia coli and may be used for routine cultivation of not particularly fastidious microorganisms.

Preparation Note

Suspend 20 g in 1000 ml distilled water. Heat to dissolve the medium completely. Sterilize by autoclaving at 15 lbs pressure (121°C) for 15 minutes. Dispense as desired.

Storage and Stability

  • Store prepared media below 8°C, protected from direct light.
  • Store dehydrated powder, in a dry place, in tightly sealed containers at 2-25°C.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service