Skip to Content
Merck
All Photos(1)

Key Documents

05-6210

Sigma-Aldrich

Copper(II) sulfate pentahydrate

JIS special grade, ≥99.5%

Synonym(s):

Cupric sulfate pentahydrate, Sulfuric acid, copper(II) salt (1:1) pentahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CuSO4 · 5H2O
CAS Number:
Molecular Weight:
249.69
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

JIS special grade

vapor pressure

7.3 mmHg ( 25 °C)

Assay

≥99.5%

form

crystalline

reaction suitability

reagent type: catalyst
core: copper

availability

available only in Japan

pH

3.7-4.5 (25 °C, 50 g/L)

mp

110 °C (dec.) (lit.)

SMILES string

O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O

InChI

1S/Cu.H2O4S.5H2O/c;1-5(2,3)4;;;;;/h;(H2,1,2,3,4);5*1H2/q+2;;;;;;/p-2

InChI key

JZCCFEFSEZPSOG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Priscilla T Y Leung et al.
Marine pollution bulletin, 124(2), 938-945 (2017-04-04)
A 96-h exposure experiment was conducted to elucidate the toxicity responses of the marine diatom Thalassiosira pseudonana upon exposure to different temperatures and copper (Cu) concentrations. Three Cu treatments (seawater control; 200μg/L Cu, EC
Bhagwan S Batule et al.
International journal of nanomedicine, 10 Spec Iss, 137-142 (2015-09-09)
We developed a simple but efficient method to synthesize protein-inorganic hybrid nanostructures with a flower-like shape (nanoflowers), which relies on sonication to facilitate the synthesis of the nanoflowers. With this technique, we synthesized nanoflowers containing laccase as a model protein
Yan Wang et al.
Pharmaceutical research, 31(11), 3106-3119 (2014-05-23)
Although Cu complexes have been investigated as anticancer agents, there has been no description of Cu itself as a cancer killing agent. A stealth liposomal Cu formulation (LpCu) was studied in vitro and in vivo. LpCu was evaluated in prostate
Fange Liu et al.
The Journal of biological chemistry, 290(25), 15621-15634 (2015-04-29)
The rubredoxin motif is present in over 74,000 protein sequences and 2,000 structures, but few have known functions. A secondary, non-catalytic, rubredoxin-like iron site is conserved in 3-hydroxyanthranilate 3,4-dioxygenase (HAO), from single cellular sources but not multicellular sources. Through the
Florence Haudin et al.
Physical chemistry chemical physics : PCCP, 17(19), 12804-12811 (2015-04-25)
The growth of chemical gardens is studied experimentally in a horizontal confined geometry when a solution of metallic salt is injected into an alkaline solution at a fixed flow rate. Various precipitate patterns are observed-spirals, flowers, worms or filaments-depending on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service