Skip to Content
Merck
All Photos(1)

Key Documents

11742027001

Roche

α-Glucosidase Assay

suitable for activity assay, kit of 1 (5 components)

Synonym(s):

α-Glucosidase, Alpha-glucosidase activity assay kit, Maltase, glucosidase, glycosidase, neutral alpha-glucosidase (NAG)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503

usage

sufficient for 30 tests (and 10 blanks)

species reactivity

human

packaging

kit of 1 (5 components)

manufacturer/tradename

Roche

storage condition

(Keep container tightly closed in a dry and well-ventilated place.)

technique(s)

activity assay: suitable

detection method

colorimetric

storage temp.

2-8°C

Related Categories

General description

Colorimetric assay for the quantitative determination of α-glucosidase (maltase) in human semen research samples. α-Glucosidase is a key enzyme involved in carbohydrate hydrolysis. It is essential for the conversion of oligosaccharides and disaccharides into glucose.

Application

α-Glucosidase Assay Kit has been used to measure neutral α-glucosidase (NAG) activity in seminal samples for the evaluation of epididymal function.

Features and Benefits

  • Fast and sensitive: The analysis can be performed using only 15 μl of semen or seminal plasma, ensuring accurate results while minimizing the sample volume needed.
  • Specific: The assay is designed to measure the activity of neutral α-glucosidase in human semen samples in life science research applications. It is not designed for diagnostic applications and cannot be used to measure α-glucosidase in other specimens.
  • Compatible: The assay is compatible with microplate assay formats and standard cuvette formats

Packaging

1 kit containing 5 components.

Components

The α-Glucosidase Assay contains the substrate and all buffers for the measurement of α-glucosidase in human semen or seminal plasma samples. It also contains a standard substance to set up a standard curve.

Preparation Note

Storage conditions (working solution): For stability of reconstituted kit components see section "General considerations".

Other Notes

For life science research only. Not for use in diagnostic procedures.

Kit Components Only

Product No.
Description

  • Reaction Buffer

  • Substrate Solution containing 4-nitrophenyl-α-D-glucopyranoside

  • Stopping Buffer

  • Castanospermine

  • Standard Solution containing 4-nitrophenol

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fatima Kazue Okada et al.
Reproduction & fertility, 1(1), 1-10 (2020-10-09)
According to the World Health Organization guidelines, ejaculatory abstinence (EA) of 2-7 days is recommended for semen analysis. This study aimed to determine how seminal quality may be affected by two EA periods from the same man. Seminal samples from
Emerging strategies for the activity assay and inhibitor screening of alpha-glucosidase
Zhang X, et al.
Food & Function, 11(1), 66-82 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service