Key Documents
ZFC0NDCA1
Milli-Q
Feed Water Conductivity Cell
For use with Milli-Q® IQ 7000, To monitor the ionic quality of feed water
About This Item
UNSPSC Code:
41104200
NACRES:
JA.13
feature
measures the conductivity of water
packaging
pkg of 1 unit
manufacturer/tradename
Milli-Q®
compatibility
for use with Milli-Q® IQ 7000
General description
The feed water conductivity cell is an installation accessory which triggers an alarm if the feed water quality is out of specifications. This prevents premature exhaustion of IPAK Meta® and IPAK Quanta® cartridges.
Application
The feed water conductivity cell is used to monitor the ionic quality of feed water for Milli-Q® IQ 7000 system.
Legal Information
IPAK META is a registered trademark of Merck KGaA, Darmstadt, Germany
IPAK QUANTA is a registered trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany