Skip to Content
Merck
All Photos(1)

Key Documents

SX0070

Supelco

Sand

Standard

Synonym(s):

Sand

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
O2Si
CAS Number:
Molecular Weight:
60.08
MDL number:
UNSPSC Code:
11101503
EC Index Number:
238-878-4

form

granular

technique(s)

ion chromatography: suitable

color

beige

particle size

0.2-0.8 mm

pH

5-8 (20 °C, 400 g/L in H2O)

bp

>2200 °C/1013 hPa

mp

1713 °C

shipped in

ambient

storage temp.

room temp

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Analysis Note

Color: Beige
Description: To pass test
Form: Granules

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service