Skip to Content
Merck
All Photos(1)

Key Documents

NI04

Sigma-Aldrich

Normal Rat IgG

This Normal Rat IgG is validated for use in ELISA Flow Cytometry Immunoblotting Immunofluorescence Immunohistochemistry Immunoprecipitation for the detection of Rat IgG, Normal.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.43

antibody form

serum

Quality Level

form

liquid

contains

≤0.1% sodium azide as preservative

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

shipped in

wet ice

storage temp.

2-8°C

General description

Native IgG from rat serum. IgG is a glycoprotein found in serum and tissue fluids. For use as a negative control in parallel with specific rat IgG-type primary antibodies.

Application

ELISA

Flow Cytometry

Immunoblotting

Immunofluorescence

Immunohistochemistry

Immunoprecipitation

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 50 mM sodium phosphate buffer, 0.2% gelatin, pH 7.5.

Other Notes

Normal IgG are provided to be used as negative controls for purified antibodies from Calbiochem. For use as a negative control, perform experiments in parallel using primary antibody or normal serum at the same concentration. A signal obtained with the normal serum indicates a nonspecific interaction; any additional signal with the primary antibody indicates a specific antibody-antigen interaction.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Adam O Whelan et al.
PloS one, 19(5), e0294998-e0294998 (2024-05-07)
Tularemia is a zoonotic disease caused by the facultative intracellular gram-negative bacterium Francisella tularensis. F. tularensis has a very low infection dose by the aerosol route which can result in an acute, and potentially lethal, infection in humans. Consequently, it
Yanchao Mu et al.
Autophagy, 14(4), 654-670 (2017-11-14)
In the advanced stages of cancer, autophagy is thought to promote tumor progression through its ability to mitigate various cellular stresses. However, the details of how autophagy is homeostatically regulated in such tumors are unknown. Here, we report that NUPR1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service