Skip to Content
Merck
All Photos(2)

Key Documents

MABT1322

Sigma-Aldrich

Anti-GCP2 Antibody, clone GCP2-02

clone GCP2-02, from mouse

Synonym(s):

Gamma-tubulin complex component 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

GCP2-02, monoclonal

species reactivity

human, frog, mouse, chicken, fish

packaging

antibody small pack of 25 μg

technique(s)

ELISA: suitable
electron microscopy: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... TUBGCP2(10844)
mouse ... Tubgcp2(74237)

General description

Gamma-tubulin complex component 2 (UniProt: Q921G8; also known as GCP2) is encoded by the Tubgcp2 (also known as Gcp2) gene (Gene ID: 74237) in murine species. Gamma-tubulin complex that is essential for microtubule nucleation at the centrosome is composed of gamma-tubulin, GCP2, GCP3, GCP4, GCP5 and GCP6. GCPs are characterized by sequence homology in two specific regions known as the grip1 and grip2 motifs. GPCs associate with gamma-tubulin to form a V-shaped subcomplex known as gamma-tubulin small complex. In this complex GCPs 2 and 3 interact via their N-terminal domain and form the arms of the V. The two-distal part of the V (C-terminal domains) bind to gamma-tubulin. The gamma-tubulin complex serves as a template for polarized growth and regulation of microtubules in eukaryotes. Significantly higher expression of both GPC2 and 3 have been reported in glioblastoma cells and they are concentrated I the centrosomes in interphase glioblastoma cells. Depletion of GPC2 and 3 causes accumulation of cells in G2/M and result in mitotic delay. (Ref.: Farache, D., et al. (2016). J. Biol. Chem. 291(44); 23112-25; Draberova, E., et al. (2015). J. Neuropathol. Exp. Neurol. 74(7); 723-42).

Specificity

Clone GCP2-02 specifically detects GCP2 in T98G glioblastoma cells. It targets an epitope with in 12 amino acids from the N-terminal region.

Immunogen

GST-tagged recombinant fragment corresponding to the first 194 amino acids from the N-terminal region of murine Gamma-tubulin complex component 2.

Application

Anti-GCP2, clone GCP2-02, Cat. No. MABT1322, is a mouse monoclonal antibody that detects Gamma-tubulin complex component 2 (CGP2) and has been tested for use in ELISA, Electron Microscopy, Immunocytochemistry, Immunoprecipitation, and Western Blotting.
Research Category
Cell Structure
Western Blotting Analysis: 1 µg/mL from a representative lot detected GCP2 in human glioblastoma T98G cell lysate.

Immunoprecipitation Analysis: A representative lot immunoprecipitated GCP2 in Immunoprecipitation applications (Draberova, E., et. al. (2015). J Neuropathol Exp Neurol. 74(7):723-42).

Immunocytochemistry Analysis: A representative lot detected GCP2 in Immunocytochemistry applications (Draberova, E., et. al. (2015). J Neuropathol Exp Neurol. 74(7):723-42).

Western Blotting Analysis: A representative lot detected GCP2 in Western Blotting applications (Draberova, E., et. al. (2015). J Neuropathol Exp Neurol. 74(7):723-42).

ELISA Analysis: A representative lot detected GCP2 in ELISA applications (Draberova, E., et. al. (2015). J Neuropathol Exp Neurol. 74(7):723-42).

Electron Microscopy Analysis: A representative lot detected GCP2 in Electron Microscopy applications (Draberova, E., et. al. (2015). J Neuropathol Exp Neurol. 74(7):723-42).

Immunoprecipitation Analysis: A represnetative lot immunoprecipitated GCP2 in mouse bone-marrow mast cells (Courtesy of Pavel Draber, Ph.D., Institute of Molecular Genetics of the ASCR, v.v.i., Prague, Czech Republic).

Quality

Evaluated by Western Blotting in T98G human glioblastoma cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected GCP2 in lysate from T98G human glioblastoma cells.

Target description

~100 kDa observed; 103.22 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service