Skip to Content
Merck
All Photos(1)

Key Documents

MAB5258A4

Sigma-Aldrich

Anti-Synaptophysin Antibody, clone SY38, Alexa Fluor 488 Conjugate

clone SY38, from mouse, ALEXA FLUOR 488

Synonym(s):

Synaptophysin, Major synaptic vesicle protein p38

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

ALEXA FLUOR 488

antibody form

purified antibody

antibody product type

primary antibodies

clone

SY38, monoclonal

species reactivity

mouse, human

species reactivity (predicted by homology)

bovine (immunogen homology)

technique(s)

immunohistochemistry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SYP(6855)

General description

Synaptophysin is a synaptic vesicle glycoprotein with four transmembrane domains weighing 38 kDa. It is present in neuroendocrine cells and in virtually all neurons in the brain and spinal cord that participate in synaptic transmission. The localization and ubiquitous expression of this protein in neuronal tissue makes it an excellent marker for neurons and neuroendocrine tumors. The gene for this protein is located on the X chromosome (Xp11.23-p11.22). The exact function of the protein is unknown: it interacts with the essential synaptic vesicle protein synaptobrevin, but when the synaptophysin gene is experimentally inactivated in animals, they still develop and function normally.

Specificity

This antibody reacts with presynaptic vesicles of cerebral and spinal neurons, of neuromuscular endplates and with the retina of man, cow, rat and mouse. It also reacts with vesicles of adrenal medulla and islet cells.

Immunogen

Vesicular fraction of bovine brain

Application

Anti-Synaptophysin Antibody, clone SY38, Alexa Fluor 488 Conjugate detects level of Synaptophysin & has been published & validated for use in IH.
Research Category
Neuroscience
Research Sub Category
Neuronal & Glial Markers

Quality

Evaluated by Immunohistochemistry in adult mouse brain tissue.

Immunohistochemistry Analysis: A 1:100 dilution of this antibody detected Synaptophysin in adult mouse brain tissue.

Target description

The unconjugated parent antibody (Cat. No. MAB5258) has an observed molecular weight at ~38 kDa

Physical form

Protein G purified
Purified mouse monoclonal IgG1κ conjugated to Alexa Fluor 488 in buffer containing 15 mg/ml BSA with 0.1% sodium azide.

Storage and Stability

Maintain refrigerated at 2-8 °C protected from light in undiluted aliquots for up to 6 months from date of receipt.

Analysis Note

Control
Adult mouse brain tissue

Other Notes

Uncongugated antibody (MAB5258) is shown to work on Rat.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daehwan Kim et al.
Translational neurodegeneration, 11(1), 16-16 (2022-03-12)
Amyotrophic lateral sclerosis (ALS) is characterized by a progressive loss of motor neurons (MNs), leading to paralysis, respiratory failure and death within 2-5 years of diagnosis. The exact mechanisms of sporadic ALS, which comprises 90% of all cases, remain unknown. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service