Skip to Content
Merck
All Photos(1)

Key Documents

AB5042P

Sigma-Aldrich

Anti-Choline Acetyltransferase Antibody

Chemicon®, from rabbit

Synonym(s):

ChAT

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

Specificity

Choline acetyltranferase. The antibody will stain cholinergic neurons in the rat central and peripheral nervous systems.

Immunogen

A 22 amino acid synthetic peptide from porcine ChAT, coupled to KLH. The peptide sequence is GLFSSYRLPGHTQDTLVAQKSS. Due to sequence similarity, the antibody is expected to react with pig, mouse primate and human ChAT.

Application

Detect Choline Acetyltransferase using this Anti-Choline Acetyltransferase Antibody validated for use in ELISA, IH.
Immunohistochemistry: 1:2,000-1:4,000 in rat basal forebrain.

Optimal working dilutions must be determined by the end user.

Physical form

Affinity purified immunoglobulin. Lyophilized. Contains no preservative. Reconstitute with 20 μL of sterile distilled water. Centrifure to remove any insoluble material.

Storage and Stability

Maintain lyophilized material dry at -70°C for up to 12 months after date of receipt. After reconstitution, maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles. Glycerol (1:1) can be added for additional stability.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service