Skip to Content
Merck
All Photos(1)

Key Documents

8.41505

Sigma-Aldrich

L-Norvaline

for synthesis

Synonym(s):

L-Norvaline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H11NO2
CAS Number:
Molecular Weight:
117.15
MDL number:
UNSPSC Code:
12352106
EC Index Number:
229-543-3
NACRES:
NA.22

Quality Level

form

crystals

mp

>310 °C

solubility

48.7 g/L

storage temp.

2-30°C

InChI

1S/C5H11NO2/c1-2-3-4(6)5(7)8/h4H,2-3,6H2,1H3,(H,7,8)

InChI key

SNDPXSYFESPGGJ-UHFFFAOYSA-N

Related Categories

Analysis Note

Assay (HClO₄): ≥ 98.0 %
Spec. rotation [α²0/D (c=7 in HCl 20%ig): +23.0 - +26.0 °
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service