Skip to Content
Merck
All Photos(1)

Key Documents

555558

Sigma-Aldrich

Rho/SRF Pathway Inhibitor, CCG-1423

The Rho/SRF Pathway Inhibitor, CCG-1423, also referenced under CAS 285986-88-1, controls the biological activity of Rho/SRF. This small molecule/inhibitor is primarily used for Cell Structure applications.

Synonym(s):

Rho/SRF Pathway Inhibitor, CCG-1423, N-(2-(4-Chloroanilino)-1-methyl-2-oxoethoxy)-3,5-bis(trifluoromethyl)benzamide, MICAL-2 Inhibitor I, MRTF-A/SRF Transcription Inhibitor I, MLK1/SRF Transcription Inhibitor I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H13ClF6N2O3
CAS Number:
Molecular Weight:
454.75
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C18H13ClF6N2O3/c1-9(15(28)26-14-4-2-13(19)3-5-14)30-27-16(29)10-6-11(17(20,21)22)8-12(7-10)18(23,24)25/h2-9H,1H3,(H,26,28)(H,27,29)

InChI key

DSMXVSGJIDFLKP-UHFFFAOYSA-N

General description

A cell-permeable benzamide compound that acts as an effective inhibitor against RhoA- and RhoC-mediated cellular activities by targeting signaling events downstream of Gα12/13 and RhoA/C, affecting MKL recruitment and/or postrecruitment function of MKL1, but not SRF-SRE interaction or ROCK kinase activity. CCG-1423 is shown to inhibit RhoC-dependent cell growth in A375M2 and SK-Mel-147 (IC50 ≤300 nM) and block RhoC/ Gα12-mediated PC-13 invasion in Matrigel assays (by ~90% at 3 µM). Shown to inhibit MICAL-2 with a Ki of 1.5 µM.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Lundquist, M.R., et al. 2014. Cell156, 563..
Evelyn, C.R., et al. 2007. Mol. Cancer Ther.6, 2249.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jitendriya Swain et al.
iScience, 26(8), 107384-107384 (2023-08-11)
Our study focused on deciphering the role of F-actin and related regulatory factors during SARS-CoV-2 particle production and transmission in human pulmonary cells. Quantitative high-resolution microscopies revealed that the late phases of SARS-CoV-2 infection induce a strong rearrangement of F-actin
Ratklao Siriwach et al.
iScience, 25(4), 104130-104130 (2022-04-09)
Keratinocyte differentiation is an intricate process that is regulated by multiple mediators. Using cultured human keratinocytes, we found that lysophosphatidic acid (LPA) induced the differentiation of a previously unsuspected keratinocyte subpopulation expressing the extracellular matrix protein, thrombospondin-1 (THBS1). This action

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service