Skip to Content
Merck
All Photos(1)

Key Documents

1.93645

Supelco

Magnesium sulfate heptahydrate

EMPLURA®

Synonym(s):

Magnesium sulfate heptahydrate, Epsom salts

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgSO4 · 7H2O
CAS Number:
Molecular Weight:
246.47
MDL number:
UNSPSC Code:
12352302

vapor density

<0.01 (vs air)

vapor pressure

<0.1 mmHg ( 20 °C)

product line

EMPLURA®

storage temp.

room temp

SMILES string

O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O

InChI

1S/Mg.H2O4S.7H2O/c;1-5(2,3)4;;;;;;;/h;(H2,1,2,3,4);7*1H2/q+2;;;;;;;;/p-2

InChI key

WRUGWIBCXHJTDG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service