Skip to Content
Merck
All Photos(2)

Key Documents

123855

Sigma-Aldrich

AKR1C3 Inhibitor

The AKR1C3 Inhibitor controls the biological activity of AKR1C3.

Synonym(s):

AKR1C3 Inhibitor, 3-(4-(Trifluoromethyl)phenylamino)benzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10F3NO2
Molecular Weight:
281.23
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥99% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

O=C(O)C1=CC(NC2=CC=C(C(F)(F)F)C=C2)=CC=C1

General description

A cell-permeable 3-phenyl-aminobenzoate compound that acts as a potent, competitive, reversible, active-site targeting inhibitor of aldo-keto reductase 1C3 (AKR1C3; type 5 17β-hydroxysteroid dehydrogenase) (IC50 = 60 nM) with excellent selectivity over other closely related human AKR isoforms (IC50 = 22.7, 15.4, 62.7, >50, and >50 µM for and AKR1C1, AKR1C2, AKR1C4, AKR1B1 and AKR1B10, respectively. Exhibits a weak inhibitory effect on cyclooxygenase 1 and 2 (IC50 = 100 µM). Shown to efficiently block the AKR1C3-mediated production of testosterone in LNCaP-AKR1C3 cells (~10 µM).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Adeniji, A.O., et al. 2012. J. Med. Chem.55, 2311.
Adeniji, A.O., et al. 2011. Bioorg. Med. Chem. Lett.21,1464.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service