Skip to Content
Merck
All Photos(1)

Key Documents

1.17276

Millipore

Mounting Kit

To upgrade of standard MAS-100 NT® and MAS-100 NT® Ex samplers with HEPA H13 filter

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103403

Agency

non-compendial

description

starter kit

feature

filter

manufacturer/tradename

MAS-100 NT®

application(s)

air monitoring
cosmetics
food and beverages
pharmaceutical

compatibility

configured for MAS-100 NT® Ex
configured for MAS-100 NT®

General description

The Mounting Kit for Filter allows an upgrade of standard MAS-100 NT® and MAS-100 NT® Ex microbial air samplers with a HEPA H13 filter. If the air samplers are used in different cleanroom classes, there is a minor risk that particles may be carried over inside an instrument. In this case, an upgrade with a HEPA H13 particle filter at the exhaust prevents any potential transfer of particles from higher to lower contaminated areas. The filtration efficiency of 99.95% has been verified under worst case conditions and is maintained even after long term usage of 1 year under normal sampling conditions (10 samples of 1000 Liter per day). No significant impact on the calibration of the instrument is observed in the study. The kit can easily be mounted to the exhaust of the air sampler by a view steps.To ease the workflow, it is recommended to combine the implementation of the mounting kit for filter with the yearly calibration/adjustment of the air sampler. Furthermore it is recommended to replace the filter itself on a yearly basis by the HEPA H13 filter 74 mm for MAS-100 NT®/MAS-100 NT® Ex w. filter.

Application

The Mounting Kit for Filter is used with MAS-100 NT® or MAS-100 NT® Ex prevents contamination transfer from higher to low contaminated areas when performing microbial air sampling of ambient air. Microbial air sampling devices are extensively used in Pharmaceutical applications. Additionally, a regulation for compounding pharmacies, USP797, that requires active air sampling in the level 5 and above areas.

Components

Filter adapter including screws, HEPA H13 filter (74 mm), allen key

Legal Information

MAS-100 NT is a registered trademark of MBV AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service