Skip to Content
Merck
All Photos(1)

Key Documents

1.11699

Millipore

Differential reinforced clostridial broth (DRCM)

GranuCult® plus, suitable for microbiology, for Clostridium spp.

Synonym(s):

Clostridium differential broth, DRCM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

Agency

DIN Norm 38411

Quality Level

sterility

non-sterile

form

granular

packaging

pkg of 500 g

manufacturer/tradename

GranuCult® plus

storage condition

dry media (Keep tightly closed)

technique(s)

microbiological culture: suitable

color

brown

pH

7.0-7.2 (25 °C, 28 g/L in H2O, after autoclaving)

solubility

27.5 g/L

bulk density

650 kg/m3

application(s)

food and beverages
microbiology

storage temp.

15-25°C

suitability

Clostridium spp.

General description

Differential Reinforced Clostridial Broth is used to determine the count of sulfite-reducing bacteria by the MPN technique. The redox indicator resazurin is used to monitor anaerobiosis. Clostridia reduce sulfite to sulfide, which causes the culture medium to turn black. As other bacteria can also produce sulfide, vegetative forms must first be removed from the culture by a relevant treatment (e.g., pasteurization), and the anaerobic spore-forming microorganisms must then be identified. The addition of polymyxin (70 IU/ml) to the broth inhibits the growth of most non-spore-forming microorganisms.

Application

Differential reinforced clostridial broth is suitable for the enumeration of all clostridia by the MPN method in foodstuffs and other materials.

Features and Benefits

  • GranuCult® offers superior granulated culture media
  • Safe and sustainable due to reduced risks associated with fine dust and toxic substance inhalation, resulting in a safer work environment
  • Excellent wettability, solubility, and free-flowing properties
  • Convenient, with minimal component separation and clumping, even under warm or humid conditions
  • High batch-to-batch reproducibility
  • Prolonged shelf life of up to five years
  • High number of test strains exceeding all regulatory demands
  • Granulation technology allows many supplements to be included, with no need to add these separately

Analysis Note

Appearance (clearness): clear
Appearance (colour): reddish-brown
pH-value (25 °C): 6.9 - 7.3
Growth (Escherichia coli ATCC 25922 (WDCM 00013)): good to very good
Growth (Bacillus cereus ATCC 11778 (WDCM 00001)): fair to good
Growth (Pseudomonas aeruginosa ATCC 27853 (WDCM 00025)): poor to fair
Growth (Clostridium bifermentans ATCC 19299): good to very good
Growth (Clostridium perfringens ATCC 13124 (WDCM 00007)): good to very good
Growth (Clostridium perfringens ATCC 10543): good to very good
Growth (Clostridium sporogenes ATCC 11437): good to very good
Growth (Clostridium sporogenes ATCC 19404 (WDCM 00008)): good to very good
Blacking (Escherichia coli ATCC 25922 (WDCM 00013)): -
Blacking (Bacillus cereus ATCC 11778 (WDCM 00001)): -
Blacking (Pseudomonas aeruginosa ATCC 27853 (WDCM 00025)): -
Blacking (Clostridium bifermentans ATCC 19299): +
Blacking (Clostridium perfringens ATCC 13124 (WDCM 00007)): +
Blacking (Clostridium perfringens ATCC 10543): +
Blacking (Clostridium sporogenes ATCC 11437): +
Blacking (Clostridium sporogenes ATCC 19404 (WDCM 00008)): +
Incubation: 7 days; 30 °C; layered with paraffin.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service