Skip to Content
Merck
All Photos(2)

Key Documents

1.10077

Supelco

Lead Test Strips, colorimetric

greener alternative

20-500 mg/L (Pb), MQuant®

Synonym(s):

Lead chemical test strips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product name

Lead Test, colorimetric, 20-500 mg/L (Pb), MQuant®

description

semi-quantitative

Quality Level

product line

MQuant®

specific analyte(s)

lead

greener alternative product characteristics

Waste Prevention
Waste Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

measuring range

20-500 mg/L (Pb)

detection method

colorimetric

greener alternative category

storage temp.

15-25°C

General description

Method: Colorimetric, with test strips and reagent, 20 - 40 - 100 - 200 - 500 mg/L (Pb), MQuant®

The MQuant® Lead test includes test strips and all necessary additional reagents and is suitable in environmental analysis. Not only in all aqueous media but also on metall surfaces it can be used to distinguish very easy between plumbiferous and non-plumbiferous metals. In acidic solution lead(II) ions react with rhodizonic acid to form a red complex. The concentration of lead(II) is measured semi-quantitatively by visual comparison of the reaction zone of the test strip with the fields of a color scale. The test strips are perfect for pre-testing a sample in an economic and quick way to estimate the lead concentration and a possible dilution factor prior to using a quantitative method such as a Spectroquant® test.

Design for sustainability benefits:

We continue to demonstrate our commitment to our planet and a sustainable future by exploring ways to improve existing products in our portfolio. We have recently reviewed the current line of MQuant® test strips to identify key opportunities to minimize impact without compromising on the product performance.

This product has benefited from:

  • Test strip with reduction
  • Recycled aluminum tube
  • Digital instruction leaflet

Learn more about the initiative: Small Changes to Test Strips for Big Impact.

Linkage

Replaces: 10077-1

Legal Information

MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service