Skip to Content
Merck
All Photos(5)

Key Documents

1.06014

Supelco

Ethyl methyl ketone

(2-butanone) EMPLURA®

Synonym(s):

Ethyl methyl ketone, 2-Butanone, Methyl ethyl ketone, MEK

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O
CAS Number:
Molecular Weight:
72.11
MDL number:
UNSPSC Code:
12191502
EC Index Number:
201-159-0
NACRES:
NA.05

vapor pressure

105 hPa ( 20 °C)

Quality Level

product line

EMPLURA®

Assay

≥99% (GC)

form

liquid

autoignition temp.

514 °C

potency

3400 mg/kg LD50, oral (Rat)
>8000 mg/kg LD50, skin (Rabbit)

expl. lim.

1.8-11.5 % (v/v)

impurities

≤0.002% Free acid (as CH3COOH)
≤0.1% Water

pH

7 (20 °C)

bp

79.6 °C/1013 hPa

mp

-86 °C

transition temp

flash point -4 °C

solubility

292 g/L

density

0.805 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C4H8O/c1-3-4(2)5/h3H2,1-2H3

InChI key

ZWEHNKRNPOVVGH-UHFFFAOYSA-N

General description

EMPLURA® is our low-cost alternative to high-purity qualities. With EMPLURA we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA solvents provide adequate specifications with the most important parameters.

Application


  • A novel similarity search approach for high-performance thin-layer chromatography (HPTLC) fingerprinting of medicinal plants.: This research introduces a new similarity search technique for HPTLC, effectively enhancing the analysis of medicinal plant extracts and potentially increasing throughput in pharmaceutical applications (Hamedi et al., 2019).

  • Determination of thermodynamic properties of poly (cyclohexyl methacrylate) by inverse gas chromatography.: The study offers a comprehensive thermodynamic analysis of polymer interactions using ethyl methyl ketone as a solvent, providing essential data for materials science and engineering (Pala et al., 2014).

  • Synthesis, growth, structural, spectral, thermal, chemical etching, linear and nonlinear optical and mechanical studies of an organic single crystal 4-chloro 4-nitrostilbene (CONS): a potential NLO material.: This article discusses the synthesis and application of a nonlinear optical material using ethyl methyl ketone, contributing to advancements in optical device technology (Kalainathan et al., 2013).

  • Synthesis, growth, structural, optical, spectral, thermal and mechanical studies of 4-methoxy 4-nitrostilbene (MONS): a new organic nonlinear optical single crystal.: Explores the development of a novel organic crystal with potential in nonlinear optical applications, leveraging ethyl methyl ketone in the synthetic process (Kalainathan et al., 2012).

  • Two-dimensional thin layer chromatographic separation of phenolic compounds from Eupatorium cannabinum extracts and their antioxidant activity.: This study utilizes two-dimensional TLC, with ethyl methyl ketone as a part of the mobile phase, to analyze the phenolic content and antioxidant activity of plant extracts, highlighting its potential in bioanalytical applications (Robak et al., 2012).

Analysis Note

Purity (GC): ≥ 99.0 %
Identity (IR): conforms
Free acid (as CH₃COOH): ≤ 0.002 %
Density (d 20 °C/ 4 °C): 0.804 - 0.805
Water: ≤ 0.1 %

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

30.2 °F - closed cup

Flash Point(C)

-1 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service