Skip to Content
Merck
All Photos(1)

Key Documents

Z-021

Supelco

Ziprasidone hydrochloride solution

1 mg/mL ((as free base), Methanol:DMSO (80:20) (m/m)), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H21ClN4OS · HCl
CAS Number:
Molecular Weight:
449.40
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1 mg/mL ((as free base), Methanol:DMSO (80:20) (m/m))

format

single component solution

storage temp.

-10 to -25°C

InChI

1S/C21H21ClN4OS.ClH.H2O/c22-17-13-18-15(12-20(27)23-18)11-14(17)5-6-25-7-9-26(10-8-25)21-16-3-1-2-4-19(16)28-24-21;;/h1-4,11,13H,5-10,12H2,(H,23,27);1H;1H2

InChI key

ZCBZSCBNOOIHFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A Certified Spiking Solution® applicable to use in forensic drug analysis or calibrator preparation by LC/MS or GC/MS. Ziprasidone is sold under the trade names Geodon and Zeldox for treatment of schizophrenia and bipolar disorder. Recent studies have suggested that Ziprasidone may increase the risk of Type II diabetes and hyperglycemia.

Application


  • Enhanced Drug Delivery Systems: A study detailed the development of sustained-release ziprasiDonemicroparticles using Soluplus and ethyl cellulose. This formulation aims to eliminate the food effect and enhance bioavailability, marking a significant advancement in pharmaceutical formulations for schizophrenia treatment (Liu W et al., 2019).

  • Targeted CNS Delivery:The study evaluated the brain-targeting and antipsychotic activities of nasally administered ziprasiDonethrough lipid-polymer hybrid nanocarriers. This novel delivery method could revolutionize treatment approaches in psychiatric medicine by enhancing drug efficacy and patient compliance in schizophrenia management (Abo El-Enin HA et al., 2023).

  • Real-world Effectiveness in Bipolar Disorder:The study conducted a register-based national cohort study on the real-world effectiveness of pharmacological treatments for bipolar disorder, including ziprasidone. This research provides valuable insights into the therapeutic impacts and side effect profiles of ziprasiDonein a large patient population (Lahteenvuo M et al., 2023).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

71.6 °F

Flash Point(C)

22 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service