Skip to Content
Merck
All Photos(1)

Key Documents

CLS33950LC

Corning® PYREX® filter discs 46 μm pore size

body diam. 45 mm, porosity 40-60 μm

Synonym(s):

Corning® 33950

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
NACRES:
NB.43

product name

Pyrex® fritted disc, body diam. 45 mm, porosity 40-60 μm

manufacturer/tradename

Corning 33950-LC

body diam.

45 mm

pore size

40-60 μm porosity

Looking for similar products? Visit Product Comparison Guide

General description

Designed for use in Pyrex brand extractors, where it is desirable to weigh the material extracted or where paper thimbles may be attacked by the chemicals used.

Legal Information

Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service