Skip to Content
Merck
All Photos(1)

Key Documents

86888

Sigma-Aldrich

Tetrabutylammonium trifluoromethanesulfonate

≥99.0% (T)

Synonym(s):

Tetrabutylammonium triflate, Trifluoromethanesulfonic acid tetrabutylammonium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)4N(CF3SO3)
CAS Number:
Molecular Weight:
391.53
Beilstein:
4172978
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99.0% (T)

form

crystals

mp

110-114 °C
112-113 °C (lit.)

SMILES string

[O-]S(=O)(=O)C(F)(F)F.CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.CHF3O3S/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;2-1(3,4)8(5,6)7/h5-16H2,1-4H3;(H,5,6,7)/q+1;/p-1

InChI key

YNJQKNVVBBIPBA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

  • Anion-capped metallohost for guest exchange: A study on the controlled release and uptake of guests in metallohosts, which used tetrabutylammonium trifluoromethanesulfonate as a precursor.(Sakata et al., 2017).
  • Deep eutectic solvents: Research on the properties of deep eutectic solvents formed by tetrabutylammonium trifluoromethanesulfonate and succinonitrile, exploring their potential applications (Yang et al., 2019).
  • Electrochemical studies: Investigation of the electrochemical reduction of CO2 in ionic liquid/organic solvent mixtures using tetrabutylammonium trifluoromethanesulfonate at gold electrodes (Yang et al., 2016).

Other Notes

Reagent used together with NBS for the activation of thioglycosides

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K. Fukase et al.
Tetrahedron Letters, 34, 2187-2187 (1993)
Claudina X Cammack et al.
Dalton transactions (Cambridge, England : 2003), 50(3), 858-868 (2020-12-22)
Non-aqueous redox flow batteries (RFBs) offer the possibility of higher voltage and a wider working temperature range than their aqueous counterpart. Here, we optimize the established 2.26 V Fe(bpy)3(BF4)2/Ni(bpy)3(BF4)2 asymmetric RFB to lessen capacity fade and improve energy efficiency over
Dmitrii Antuganov et al.
Journal of pharmaceutical and biomedical analysis, 173, 68-74 (2019-05-24)
Fluorine-18 labelled radiotracers are the most common diagnostic agents employed in positron emission tomography (PET). Despite well-established quality control (QC) procedures, introduction of new synthetic methods demands continuous development of analytical methodology. Here we propose capillary electrophoresis (CE) as a
Ken T Ngo et al.
Journal of the American Chemical Society, 139(7), 2604-2618 (2017-01-25)
Electrocatalytic reduction of CO

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service