Skip to Content
Merck
All Photos(1)

Key Documents

803170

Sigma-Aldrich

Vitamin B12-(dimethylbenzimidazole-13C7) solution

1 μg/mL in methanol, 99 atom % 13C, 95% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C7C56H88CoN14O14P
Molecular Weight:
1362.31
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99 atom % 13C

Quality Level

Assay

95% (CP)

form

solution

concentration

1 μg/mL in methanol

technique(s)

mass spectrometry (MS): suitable

color

colorless

application(s)

detection

mass shift

M+7

storage temp.

−20°C

SMILES string

[Co+]C#N.C/C(C1=N[C@]2(C)[C@](CC(N)=O)(C)[C@@H]1CCC(N)=O)=C3[C@](CC(N)=O)(C)C(CCC(N)=O)C(CC4C(C)(C)[C@H](CCC(N)=O)C(/C(C)=C5[C@](CCC(NCC(OP(O[C@H]6[C@@H](O)[C@H](N([13CH]=N7)[13C]8=[13C]7[13CH]=[13C](C)[13C](C)=[13CH]8)O[C@@H]6CO)([O-])=O)C)=O)(C)C(CC(N)=

InChI

1S/C62H90N13O14P.CN.Co/c1-29-20-39-40(21-30(29)2)75(28-70-39)57-52(84)53(41(27-76)87-57)89-90(85,86)88-31(3)26-69-49(83)18-19-59(8)37(22-46(66)80)56-62(11)61(10,25-48(68)82)36(14-17-45(65)79)51(74-62)33(5)55-60(9,24-47(67)81)34(12-15-43(63)77)38(71-55)23-42-58(6,7)35(13-16-44(64)78)50(72-42)32(4)54(59)73-56;1-2;/h20-21,28,31,34-37,41-42,52-53,57,76,84H,12-19,22-27H2,1-11H3,(H2,63,77)(H2,64,78)(H2,65,79)(H2,66,80)(H2,67,81)(H2,68,82)(H,69,83)(H,85,86);;/q;;+1/p-1/b54-32-,55-33-;;/t31?,34?,35-,36-,37?,41-,42?,52-,53-,57-,59+,60+,61+,62+;;/m1../s1/i20+1,21+1,28+1,29+1,30+1,39+1,40+1;;

InChI key

FTKDYBHYJJLMND-SDKDZEEISA-M

Application

Vitamin B12-(dimethylbenzimidazole-13C7) can be used as a stable isotope internal standard for accurate data interpretation in specific biological samples.

Packaging

1 mL in ampule
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service