Skip to Content
Merck
All Photos(1)

Key Documents

69806

Sigma-Aldrich

(+)-tert-Butyl D-lactate

≥99.0% (sum of enantiomers, GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14O3
CAS Number:
Molecular Weight:
146.18
Beilstein:
1722069
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.0% (sum of enantiomers, GC)

form

crystals

optical activity

[α]20/D +7.3±0.5°, c = 1.7% in methylene chloride

optical purity

enantiomeric ratio: ≥99.5:0.5 (GC)

mp

38-42 °C

storage temp.

2-8°C

SMILES string

C[C@@H](O)C(=O)OC(C)(C)C

InChI

1S/C7H14O3/c1-5(8)6(9)10-7(2,3)4/h5,8H,1-4H3/t5-/m1/s1

InChI key

IXXMVXXFAJGOQO-RXMQYKEDSA-N

Other Notes

Chiral building block; synthesis of depsipeptides by reaction with amino acid derivs.; Hydroxy-group activation and substitution reaction

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

U. Schmidt et al.
Synthesis, 475-475 (1988)
H.C. Nung et al.
Tetrahedron Letters, 33, 2629-2629 (1992)
H. Kunz et al.
Angewandte Chemie (International Edition in English), 84, 798-798 (1984)
Wei-He Qian et al.
International journal of molecular medicine, 44(2), 569-581 (2019-06-08)
Alcoholic hepatitis (AH) is a fatal inflammatory syndrome with no effective treatments. Intestinal injury and intestinal endotoxemia (IETM) contribute greatly in the development of AH. MicroRNAs (miRNAs/miRs) have been reported to affect intestinal injury. The present study aims to investigate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service