Skip to Content
Merck
All Photos(1)

Key Documents

662720

Sigma-Aldrich

3-Bromo-2-methylpyridine

97%

Synonym(s):

3-Bromo-2-picoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6BrN
CAS Number:
Molecular Weight:
172.02
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

refractive index

n20/D 1.5604

density

1.495 g/mL at 25 °C

SMILES string

Cc1ncccc1Br

InChI

1S/C6H6BrN/c1-5-6(7)3-2-4-8-5/h2-4H,1H3

InChI key

AIPWPTPHMIYYOX-UHFFFAOYSA-N

Application

Conversion to the corresponding pyridine carboxaldehyde via peroxide initiated NBS gem-dibromination followed by hydrolysis.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

174.0 °F - closed cup

Flash Point(C)

78.9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 46, 6033-6033 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service