Skip to Content
Merck
All Photos(2)

Key Documents

655597

Sigma-Aldrich

Methyl 4-(cyanomethyl)benzoate

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OOCC6H4CH2CN
CAS Number:
Molecular Weight:
175.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

bp

110 °C/0.2 mmHg (lit.)

mp

55-58 °C (lit.)

SMILES string

COC(=O)c1ccc(CC#N)cc1

InChI

1S/C10H9NO2/c1-13-10(12)9-4-2-8(3-5-9)6-7-11/h2-5H,6H2,1H3

InChI key

XRZGMNGGCZTNGE-UHFFFAOYSA-N

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service