Skip to Content
Merck
All Photos(1)

Key Documents

615390

Sigma-Aldrich

2-Ketoglutaric acid-d6

98 atom % D, 99% (CP)

Synonym(s):

α-Ketoglutaric acid-d6, α-Ketopentanoic acid-d6, 2-Ketopentanedioic acid-d6, 2-Oxoglutaric acid-d6

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
DO2CCD2CD2COCO2D
CAS Number:
Molecular Weight:
152.14
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:

isotopic purity

98 atom % D

Assay

99% (CP)

form

solid

mp

114-117 °C

mass shift

M+6

SMILES string

[2H]OC(=O)C(=O)C([2H])([2H])C([2H])([2H])C(=O)O[2H]

InChI

1S/C5H6O5/c6-3(5(9)10)1-2-4(7)8/h1-2H2,(H,7,8)(H,9,10)/i1D2,2D2/hD2

InChI key

KPGXRSRHYNQIFN-DTDGFSOZSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service