Skip to Content
Merck
All Photos(2)

Key Documents

591130

Sigma-Aldrich

3-Methoxycarbonylphenylboronic acid

Synonym(s):

Methyl 3-boronobenzoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3O2CC6H4B(OH)2
CAS Number:
Molecular Weight:
179.97
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

mp

205-208 °C (lit.)

SMILES string

COC(=O)c1cccc(c1)B(O)O

InChI

1S/C8H9BO4/c1-13-8(10)6-3-2-4-7(5-6)9(11)12/h2-5,11-12H,1H3

InChI key

ALTLCJHSJMGSLT-UHFFFAOYSA-N

Application

Reactant involved in:
  • Suzuki-Miyaura cross-coupling
  • Iterative cross-coupling of boronate building blocks
  • Cross-coupling with aryl / alkenyl sulfonates
  • Synthesis of symmetrical biaryls via CuCl catalyzed homocoupling
  • Trifluoromethylation
  • Cyanation

Other Notes

Contains varying amounts of anhydride

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service