Skip to Content
Merck
All Photos(1)

Key Documents

547247

Sigma-Aldrich

N-Boc-4-piperidineethanol

97%

Synonym(s):

N-(tert-Butoxycarbonyl)-4-piperidineethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H23NO3
CAS Number:
Molecular Weight:
229.32
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.4730 (lit.)

bp

120-150 °C/0.5 mmHg (lit.)

density

1.043 g/mL at 25 °C (lit.)

SMILES string

CC(C)(C)OC(=O)N1CCC(CCO)CC1

InChI

1S/C12H23NO3/c1-12(2,3)16-11(15)13-7-4-10(5-8-13)6-9-14/h10,14H,4-9H2,1-3H3

InChI key

YBNJZIDYXCGAPX-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service