Skip to Content
Merck
All Photos(2)

Key Documents

538329

Sigma-Aldrich

Acetoacetanilide

≥99.5%

Synonym(s):

1-(Phenylamino)-1,3-butanedione, 1-(Phenylcarbamoyl)-2-propanone, 3-Oxo-N-phenylbutanamide, 3-Oxo-N-phenylbutyramide, 4-(Phenylamino)-2,4-butanedione, Acetoacetamidobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH2CONHC6H5
CAS Number:
Molecular Weight:
177.20
Beilstein:
473419
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

6.1 (vs air)

Assay

≥99.5%

autoignition temp.

843 °F

mp

83-88 °C (lit.)

SMILES string

CC(=O)CC(=O)Nc1ccccc1

InChI

1S/C10H11NO2/c1-8(12)7-10(13)11-9-5-3-2-4-6-9/h2-6H,7H2,1H3,(H,11,13)

InChI key

DYRDKSSFIWVSNM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Acetoacetanilide (N-phenyl-3-oxobutanamide) undergoes condensation reaction with o-phenylenediamine to afford a Schiff base. Dielectric constant of acetoacetanilide crystals were found to increase on exposure to 120MeV Ag13+ ions due to increase in number of defects. Acetoacetanilide/Ce4+ system serves as an initiator during the polymerization of vinyl monomers.

Application

Acetoacetanilide may be used to synthesize:
  • azo pigments
  • acetoacetanilido-4-aminoantipyrine (Schiff base)
  • 6-aryl-2-methyl-4-oxo-N,N′-diphenyl-2-cyclohexene-1,3-dicarboxamides
  • photoluminescent lanthanide complexes

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and photoluminescent properties of lanthanides acetoacetanilide complexes
Souza, E. R., et al.
Journal of Fluorescence, 23.5, 939-946 (2013)
Crystal structures of azo pigments derived from acetoacetanilide
Whitaker, A.
Journal of the Society of Dyers and Colourists, 104.7-8 , 294-300 (1988)
Effect of swift heavy ion (SHI) irradiation on dielectric properties of acetoacetanilide crystals
Prabhu SG, et al.
Nuclear Instruments & Methods in Physics Research. Section B, Beam Interactions With Materials and Atoms null
Synthesis and characterisation of Cu (II), Ni (II), Mn (II), Zn (II) and VO (II) Schiff base complexes derived fromo-phenylenediamine and acetoacetanilide
Raman N, et al.
Journal of Chemical Sciences (Bangalore), 113.3 , 183-189 (2001)
Formation of 6-aryl-2-methyl-4-oxo-N, N?-diphenyl-2-cyclohexene-1, 3-dicarboxamides from acetoacetanilide and aromatic aldehydes catalyzed by a mixture of aryl amines and iodine
Gein VL, et al.
Russ. J. Gen. Chem., 86.1, 58-61 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service