Skip to Content
Merck
All Photos(2)

Key Documents

472166

Sigma-Aldrich

4-Fluorobenzenesulfonyl isocyanate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4SO2NCO
CAS Number:
Molecular Weight:
201.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

bp

63 °C/0.1 mmHg (lit.)

mp

33-38 °C (lit.)

SMILES string

Fc1ccc(cc1)S(=O)(=O)N=C=O

InChI

1S/C7H4FNO3S/c8-6-1-3-7(4-2-6)13(11,12)9-5-10/h1-4H

InChI key

FHMRJRSOWRCSKA-UHFFFAOYSA-N

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yaws CL.
Thermophysical Properties of Chemicals and Hydrocarbons, 284-284 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service