Skip to Content
Merck
All Photos(1)

Key Documents

440698

Sigma-Aldrich

Strontium isopropoxide

99.9% trace metals basis

Synonym(s):

Strontium diisopropoxide, Strontium isopropoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Sr(OCH(CH3)2)2
CAS Number:
Molecular Weight:
205.79
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9% trace metals basis

form

crystalline powder

reaction suitability

core: strontium

SMILES string

CC(C)O[Sr]OC(C)C

InChI

1S/2C3H7O.Sr/c2*1-3(2)4;/h2*3H,1-2H3;/q2*-1;+2

InChI key

OHULXNKDWPTSBI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1 - Self-heat. 2 - Skin Corr. 1B

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daxian Cao et al.
Nano letters, 20(3), 1483-1490 (2019-09-24)
All-solid-state lithium batteries (ASLBs) are promising for the next generation energy storage system with critical safety. Among various candidates, thiophosphate-based electrolytes have shown great promise because of their high ionic conductivity. However, the narrow operation voltage and poor compatibility with

Articles

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service