Skip to Content
Merck
All Photos(1)

Key Documents

39000

Sigma-Aldrich

2-Dimethylaminoethanol (+)-bitartrate salt

≥99.0%

Synonym(s):

Atrol, N,N-Dimethylethanolamine (+)-bitartrate salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NCH2CH2OH · C4H6O6
CAS Number:
Molecular Weight:
239.22
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.0%

optical activity

[α]20/D +18±1°, c = 3% in H2O

reaction suitability

reaction type: solution phase peptide synthesis

mp

109-113 °C

application(s)

peptide synthesis

SMILES string

CN(C)CCO.O[C@H]([C@@H](O)C(O)=O)C(O)=O

InChI

1S/C4H11NO.C4H6O6/c1-5(2)3-4-6;5-1(3(7)8)2(6)4(9)10/h6H,3-4H2,1-2H3;1-2,5-6H,(H,7,8)(H,9,10)/t;1-,2-/m.1/s1

InChI key

UIEGYKVRCKDVKQ-LREBCSMRSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Josie A Silvaroli et al.
ACS chemical biology, 14(3), 434-448 (2019-02-06)
Cellular retinol-binding proteins (CRBPs) facilitate the uptake and intracellular transport of vitamin A. They integrate retinoid metabolism, playing an important role in regulating the synthesis of bioactive vitamin A metabolites. Thus, CRBPs constitute potential pharmacological targets to modulate cellular retinoid
Nicolas Taveau et al.
Experimental & molecular medicine, 52(7), 1090-1101 (2020-07-10)
Environmental light has deleterious effects on the outer retina in human retinopathies, such as ABCA4-related Stargardt's disease and dry age-related macular degeneration. These effects involve carbonyl and oxidative stress, which contribute to retinal cell death and vision loss. Here, we
Ala Morshedian et al.
Neuron, 102(6), 1172-1183 (2019-05-06)
While rods in the mammalian retina regenerate rhodopsin through a well-characterized pathway in cells of the retinal pigment epithelium (RPE), cone visual pigments are thought to regenerate in part through an additional pathway in Müller cells of the neural retina.
Philipp M Amann et al.
Skin pharmacology and physiology, 28(4), 205-212 (2015-02-28)
Vitamin A (all- trans -retinol, ATRol) serves as a precursor for all- trans -retinoic acid (ATRA), a ligand for the retinoic acid receptor (RAR), representing a potent regulator for many physiological processes. While murine melanoma cells are highly sensitive to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service