Skip to Content
Merck
All Photos(2)

Key Documents

379913

Sigma-Aldrich

Hydroxylamine sulfate

99.999% trace metals basis

Synonym(s):

Hydroxylammonium sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH2OH)2 · H2SO4
CAS Number:
Molecular Weight:
164.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99.999% trace metals basis

form

solid

mp

170 °C (dec.) (lit.)

solubility

water: soluble(lit.)

SMILES string

NO.NO.OS(O)(=O)=O

InChI

1S/2H3NO.H2O4S/c2*1-2;1-5(2,3)4/h2*2H,1H2;(H2,1,2,3,4)

InChI key

VRXOQUOGDYKXFA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The direct titration of alkaline ferricyanide in presence of zinc sulfate with hydroxylamine sulfate, for the detection of hydrarine and hydroxylamine in mixture has been reported. Influence of hydroxylamine sulfate on the nuclear cycle and the induction of endoreduplication in Zea mays root tips has been investigated.

Application

Hydroxylamine sulfate may be used to prepare highly sensitive cellulose tape, used for the detection of formaldehyde gas. It may be used in the quantitative determination of perchlorate in biological fluids by spectrophotometric methods .

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Carc. 2 - Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Oral

Target Organs

Blood

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spectrophotometric determination of micro amounts of perchlorate in biological fluids.
J A Weiss et al.
Analytical chemistry, 44(3), 619-620 (1972-03-01)
Endoreduplication induced by hydroxylamine sulfate in Zea mays root tip nuclei.
M S Lin et al.
Experimental cell research, 86(1), 47-52 (1974-05-01)
Development of a monitoring tape for formaldehyde using hydroxylamine sulfate and methyl yellow.
NAKANO N, et al.
Analytical Sciences, 10(4), 641-645 (1994)
Oxidation of hydroxylamine by ferricyanide in presence of zinc sulfate: A rapid method for estimating hydroxylamine and hydrazine in a mixture.
Sant BR.
Analytica Chimica Acta, 20, 371-375 (1959)
Ji Bao et al.
Scientific reports, 5, 10756-10756 (2015-06-02)
Whole-liver perfusion-decellularization is an attractive scaffold-preparation technique for producing clinical transplantable liver tissue. However, the scaffold's poor hemocompatibility poses a major obstacle. This study was intended to improve the hemocompatibility of perfusion-decellularized porcine liver scaffold via immobilization of heparin. Heparin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service