Skip to Content
Merck
All Photos(1)

Key Documents

193097

Sigma-Aldrich

trans-3-Hexenoic acid

97%

Synonym(s):

(3E)-3-Hexenoic acid, (3E)-Hex-3-enoic acid, (E)-3-Hexenoic acid, (E)-Hex-3-enoic acid, 3(E)-Hexenoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH=CHCH2CO2H
CAS Number:
Molecular Weight:
114.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

>1 (vs air)

Assay

97%

form

liquid

refractive index

n20/D 1.44 (lit.)

bp

118-119 °C/22 mmHg (lit.)

mp

11-12 °C (lit.)

density

0.963 g/mL at 25 °C (lit.)

SMILES string

CC\C=C\CC(O)=O

InChI

1S/C6H10O2/c1-2-3-4-5-6(7)8/h3-4H,2,5H2,1H3,(H,7,8)/b4-3+

InChI key

XXHDAWYDNSXJQM-ONEGZZNKSA-N

Looking for similar products? Visit Product Comparison Guide

Application

trans-3-Hexenoic acid was used in the preparation of trans-3-hexen-1-ol, starting reagent for the synthesis of endo-brevicomin.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Improved synthesis ofendo-brevicomin for the control of bark beetles (Coleoptera: Scolytidae).
Look M.
Journal of Chemical Ecology, 2(1), 83-86 (1976)
Fangfang Zeng et al.
Insect biochemistry and molecular biology, 113, 103213-103213 (2019-08-24)
Mosquitoes rely heavily on the olfactory system to find a host for a bloodmeal, plants for a source of energy and suitable sites for oviposition. Here, we examined a cluster of eight odorant receptors (ORs), which includes one OR, CquiOR1
Pingxi Xu et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(46), 16592-16597 (2014-10-29)
Insect repellents are important prophylactic tools for travelers and populations living in endemic areas of malaria, dengue, encephalitis, and other vector-borne diseases. DEET (N,N-diethyl-3-methylbenzamide) is a 6-decade-old synthetic repellent, which is still considered the gold standard of mosquito repellents. Mosquitoes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service