Skip to Content
Merck
All Photos(2)

Key Documents

14944

Sigma-Aldrich

N-[4-(3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Heptadecafluorodecyl) benzyloxycarbonyloxy]succinimide

≥97.0% (NMR)

Synonym(s):

N-[4-(1H,1H,2H,2H-Perfluorodecyl)benzyloxycarbonyloxy]succinimide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H14F17NO5
CAS Number:
Molecular Weight:
695.32
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (NMR)

form

solid

storage temp.

−20°C

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CCc1ccc(COC(=O)ON2C(=O)CCC2=O)cc1

InChI

1S/C22H14F17NO5/c23-15(24,8-7-10-1-3-11(4-2-10)9-44-14(43)45-40-12(41)5-6-13(40)42)16(25,26)17(27,28)18(29,30)19(31,32)20(33,34)21(35,36)22(37,38)39/h1-4H,5-9H2

InChI key

DLEHONFUGNNFOG-UHFFFAOYSA-N

Legal Information

Product of Fluorous Technologies, Inc. Use of this compound may be protected by U.S. patents 6,156,896; 5,859,247; 5,777,121 and 6,673,539.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Lact. - Repr. 1B - STOT RE 1

Target Organs

Liver

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service