Merck
HomeChemical Analysis for Food and Beverage TestingLC-MS/MS Analysis of 16 PFAS in Salmon Using QuEChERS Based on FDA Method C-010.02
LC-MS/MS Analysis of 16 PFAS in Salmon Using QuEChERS Based on FDA Method C-010.02
Introduction
Per- and polyfluorinated alkyl substances (PFAS), also known as Forever Chemicals, are a class of compounds that have been widely used as commercial product applications over the past decades due to their versatile physical and chemical properties (e.g., water-repellent, firefighting foams, cookware, food packaging). Due to their chemical stability, PFAS chemicals are prevalent in our environment and pose a potential risk of bioaccumulation in humans over time. To limit the possible risks to human health, such as low infant birth weights, cancer, and impacts on the immune system, regulatory agencies such as EPA and FDA have established PFAS regulations encompassing limit values and PFAS analytical methods for specific PFAS substances.1-4 The U.S. Food and Drug Administration (FDA) issued advisories for PFAS extraction from food samples applying modified QuEChERS (Quick, Easy, Cheap, Effective, Rugged, and Safe) methodology extraction and clean-up step (using dispersive solid phase extraction - dSPE), and an additional clean-up of the obtained QuEChERS extract using weak anion exchange (WAX) SPE columns, followed by LC-MS/MS analysis.4 For FDA method C-010.024, an extraction salt mixture containing 6.0 g MgSO4 as well as 1.5 g NaCl, and a dSPE clean-up mix containing 900 mg MgSO4, 300 mg primary secondary amine (PSA) and 150 mg graphitized carbon black, are specified. The Supel™ QuE non-buffered extraction salt mix (55295-U) and the newly developed Supel™ QuE PSA/ENVI-Carb™ Tube 3 for clean-up (55479-U) meet these method requirements. For further clean-up in this study, Supelclean™ ENVI-WAX™ SPE cartridges were used as a direct equivalent to the specified SPE cartridges in the FDA method C-010.02. This application note outlines one of the PFAS detection methods, developed in accordance with the FDA method C-010.02, for analyzing 16 PFAS compounds in salmon using LC-MS/MS.
Experimental— PFAS Analysis Method
Preparation of PFAS Standards and Solutions
Native and isotopically labeled PFAS standards were obtained as methanolic 50 µg/mL stock solutions. These standards were then diluted with methanol following the dilution scheme of the method C-010.02 to obtain the calibration standards in the required concentration range (external calibration: 0.01, 0.05, 0.10, 0.50, 1.0, 5.0, 10, and 25 ng/mL).
Sample Preparation
In accordance with U.S. FDA method C-010.02, PFAS analysis was performed for salmon samples. The salmon samples (5 g, homogenized with knife mill) were spiked at 0.05, 0.5 or 2.0 ng/g with 16 native PFAS as well as further spiked with 8 isotopically labeled internal standards at 2 ng/g. After adding 5 mL water, 150 µL formic acid and 10 mL acetonitrile, the samples were vigorously shaken for PFAS extraction. To test for potential PFAS background contamination, salmon samples were only spiked with isotopically labeled internal standards and equally processed. After addition of the Supel QuE salt package (6.0 g MgSO4, 1.5 g NaCl, 55295-U), the mixture was placed on a shaker for 10 minutes at 1500 rpm, followed by centrifugation for 10 minutes at 4000 g. The obtained supernatant was transferred into a second tube, the Supel™ QuE PSA/ ENVI-Carb™ Tube 3 (contains 900 mg MgSO4, 300 mg PSA, 150 mg ENVI-Carb™, 55479-U), and the sample was shaken for 10 minutes at 1500 rpm, followed by centrifugation for 10 minutes at 4000 g. Subsequently, the supernatants were filtered using Millex® Nylon 0.2 µm syringe filters (SLGNX13). For salmon samples, further SPE clean-up is required prior to LC-MS/MS analysis in accordance with the above FDA method. Therefore, 1 mL of the filtered supernatant was transferred into a tube and diluted to 15 mL with water. SupelcleanENVI-WAX SPE cartridges were used (500 mg, 6 mL, 54057-U) for the SPE clean-up, conditioned with 9 mL of 0.3% NH4OH in MeOH and equilibrated with 5 mL of water. After the salmon sample (15 mL) was loaded and passed through the cartridge, 5 mL of water was added as a washing step. The cartridge was subsequently dried for 1 min before 4 mL 0.3% NH4OH in MeOH was used to elute the analytes into an additional tube. Finally, the sample was evaporated to dryness, reconstituted in 1 mL MeOH and 5 µL of the internal standard N-EtFOSAA-d5 (200 ng/ml) were added prior to LC-MS/MS analysis.
LC-MS/MS analysis
An Agilent 1290 Infinity II instrument coupled to an Agilent 6495C triple quadrupole mass spectrometer was used for the LC-MS/MS analysis. Analyte separation was achieved using a Purospher® Star RP-18 endcapped (2 µm) Hibar HR (15 cm x 2.1 mm, 1.50649) as analytical column. In addition, a delay column (Purospher® Star RP18 endcapped Hibar RT (5 cm x 4.0 mm, 3 µm, 1.50428) was positioned after the mixing valve and before the autosampler to offset potential PFAS contamination originating from the LC system (e.g., pump, tubings, fittings, filters). The Purospher® STAR HPLC columns were chosen, as they generally provide stable HPLC separations even at higher pH, e.g. in the presence of basic mobile phase additives, such as 1-methyl piperidine recommended to be used in the C-010.02 method. We have tested the method with and without addition of the 1-methyl piperidine to the mobile phase. As no significant sensitivity gain was obtained using the additive in the presented application, however, the separation was performed without using 1-methyl piperidine, but the column was kept for this application to be prepared also for basic MP conditions. To avoid possible PFAS adsorption to the glass surface, polypropylene snap caps vials were used instead of standard glass vials. The LC and MS conditions are listed in Table 1.
LC Conditions
Instrument:
Agilent 1290 Infinity II instrument coupled to an Agilent 6495C triple quadrupole mass spectrometer
Columns:
Purospher® STAR RP-18 endcapped, Hibar HR, 2.0 µm, 15 cm x 2.1 mm (1.50649)
Delay Column: Purospher® Star RP-18 endcapped, Hibar RT, 3.0 µm, 5 cm x 4.0 mm (1.50428)
Mobile phase:
[A] 5 mM Ammonium acetate*; [B] methanol
Gradient:
Time (min)
A%
B%
Initial
90.0
10.0
3.0
90.0
10.0
3.1
60.0
40.0
26.0
10.0
90.0
26.1
90.0
10.0
28.0
90.0
10.0
Flow rate:
0.30 mL/min
Column temp.:
40 °C
Detector:
MS (ESI-), MRM (see Table 2 for details)
Injection volume:
5 µL
*Mobile phase A was adjusted compared to FDA method C-010.02 and used without addition of 1-methyl piperidine
Table 1.LC conditions used for PFAS analysis of 16 compounds
Peak
Acronym
Compound
MRM
Collision energy
RT (min)
R2
1
PFBA
Perfluorobutanoic acid
213.0–169.0
4
7.2
0.9969
2
PFPeA
Perfluoropentanoic acid
263.0–219.0
4
10.0
0.9972
3
PFBS
Perfluorobutanesulfonic acid
298.9–80.0
40
10.6
0.9967
4
PFHxA
Perfluorohexanoic acid
313.0–269.0
4
12.9
0.9969
5
PFPeS
Perfluoropentanesulfonic acid
348.9–99.0
37
13.3
0.9962
6
HFPO-DA
Hexafluoropropylene oxide dimer acid
285.0–169.0
4
13.6
0.9969
7
PFHpA
Perfluoroheptanoic acid
363.0–319.0
4
15.5
0.9924
8
PFHxS
Perfluorohexanesulfonic acid
398.9–99.0
41
15.8
0.9935
9
NaDONA
Sodium dodecafluoro-3H-4,8-dioxanonanoate
377.0–251.0
8
15.8
0.9985
10
PFOA
Perfluoroctanoic acid
413.0–369.0
8
17.8
0.9988
11
PFHpS
Perfluoroheptanesulfonic acid
448.9–99.0
45
17.9
0.9969
12
PFNA
Perfluorononanoic acid
463.0–419.0
8
19.8
0.9973
13
PFOS
Perfluorooctanesulfonic acid
498.9–80.0
76
19.8
0.9957
14
9Cl-PF3ONS
9-Chlorohexadecafluoro-3-oxanonane-1-sulfonic acid
530.9–351.0
28
20.7
0.9899
15
PFDA
Perfluorodecanoic acid
513.0–469.0
8
21.4
0.9822
16
11Cl-PF3OUdS
11-Chloroeicosafluoro-3-oxaundecane-1-sulfonic acid
630.9–451.0
32
23.5
0.9978
Table 2.MRM, chromatographic and linearity (R2) data for 16 PFAS analytes
Results and Discussion
A chromatogram of a solvent calibration standard containing the 16 native compounds is shown in Figure 1. The lower limit of quantitation (LLOQ) for all 16 compounds was 0.01 ng/mL for the LC-MS/MS method, and 0.02 ng/g in relation to the salmon sample. Linear calibration curves (0.01 ─25 ng/mL) with R2 ≥0.99 were obtained for all PFAS analytes except for PFDA with 0.98 (Table 2).
A chromatogram depicting the relationship between response (measured in counts) on the y-axis and retention time (measured in minutes) on the x-axis. The chromatogram showcases distinct peaks designated by numbers 1 to 16, each marked with different colors. These peaks are a result of the LC-MS/MS analysis of a solvent calibration standard, wherein the 16 PFAS compounds are present at a concentration of 1 ng/mL in methanol.
Figure 1.16 PFAS compounds at 1 ng/mL in methanol (Peak ID see Table 2)
The acceptable recovery range for the investigated PFAS analytes based on the FDA guidelines for the validation of chemical methods in food, feed, cosmetics and veterinary products is 40-120% (including RSD ≤ 22%) for concentrations at 0.001 mg/kg (1 ng/g). Table 3 displays the recoveries and %RSD from the experimental study where 16 compounds were spiked in triplicate to salmon samples at concentration levels of 0.05, 0.5, and 2.0 ng/g. The background evaluation of PFAS in the used salmon (salmon sample only spiked with isotopically labeled internal standards) revealed analyte concentrations for 3 PFAS analytes present above LLOQ (0.02 ng/g for PFBA and PFOS, 0.04 ng/g for PFOA). To accurately calculate the analyte recovery at the lowest spiking level (0.05 ng/g) for PFBA, PFOS and PFOA, the background levels found in salmon were subtracted accordingly. All obtained recoveries and %RSD were in the recommended range and thus meet the requirements of the FDA method.5
Fortified conc. 0.05 ng/g
Fortified conc. 0.5 ng/g
Fortified conc. 2.0 ng/g
Analyte
Mean recovery (%)
RSD (%)
Mean recovery [%]
RSD [%]
Mean recovery [%]
RSD [%]
PFBA
88.6**
1.4
81.5
2.1
71.5
0.9
PFPeA
98.8
8.1
76.9
3.1
71.4
1.6
PFBS
97.6
1.6
77.7
3.0
71.8
3.1
PFHxA
107.3
2.0
77.6
3.1
71.2
2.2
PFPeS
97.9
3.4
80.5
3.3
69.6
4.5
HFPO-DA
92.5
3.7
75.7
4.0
72.2
1.8
PFHpA
111.7
9.6
79.2
7.7
70.1
2.5
PFHxS
104.0
10.9
85.6
4.0
75.1
3.9
NaDONA
92.6
3.4
72.8
1.9
70.2
0.2
PFOA
91.8 **
14.1
97.9
6.0
78.4
0.4
PFHpS
86.9
13.8
75.8
4.3
66.3
3.5
PFNA
99.9
13.9
96.4 (87.0 & 105.7) *
N/A
89.8
21.9
PFOS
88.4** (85.2&91.5) *
N/A
86.4
2.2
75.0
8.0
9Cl-PF3ONS
82.3
7.7
70.8
4.4
64.6
2.2
PFDA
82.6 (71.2 & 94.0) *
N/A
78.0 (73.1 & 82.8) *
N/A
82.6 (70.9 & 94.3) *
N/A
11Cl-PF3OUdS
90.7
6.6
63.0
4.1
56.0
2.1
* n = 2, due to discarding of outlier
** Analyte recovery at lowest spiking level obtained after subtraction of background level in salmon (0.02 ng/g for PFBA and PFOS, 0.04 ng/g for PFOA)
Table 3.Precision and recovery (n = 3) of PFAS in salmon samples at 3 fortification levels (0.05 ng/g, 0.5 ng/g and 2.0 ng/g)
Conclusion
In this application note, the workflow of the FDA method C-010.02 for analysis of 16 PFAS in salmon samples based on a QuEChERS approach was investigated. At fortification levels of 0.05 ng/g, 0.5 ng/g and 2.0 ng/g, recoveries for all 16 compounds were within the acceptable range specified in the relevant FDA guideline for method validation. The calculated % RSDs were below 22%, further indicating a satisfactory precision. Hence, the employed combination of the new Supel QuE PSA/ENVI-CarbTube 3 clean-up mix (55479-U), alongside the Supel QuE extraction salt mix, the Supelclean ENVI-WAX SPE cartridges, and Millex syringe filters, demonstrated efficacy as suitable sample preparation products in PFAS testing methods,  for achieving the desired clean-up of food samples. The Purospher® STAR RP-18 HPLC columns were used in this method, as these generally provide stable separations in a wider range of mobile phase pH values, e.g. in the presence of basic mobile phases additives, such as 1-methyl piperidine described in the C-010.02 method.
See also our dedicated 2.7 µm Fused-Core® particle Ascentis® Express 90 Å PFAS and the Ascentis® Express 90 Å PFAS Delay columns.
Read more about PFAS testing at SigmaAldrich.com/pfas
Related Products
HPLC Columns, Sample Prep, Reagents & Solvents
Sorry, an unexpected error has occurred
Network error: Failed to fetch
Discover our range of PFAS certified reference materials (CRMs) and analytical standards for reliable PFAS analysis and quantification. Click here to see the full list.
Certified Reference Materials (CRMs)
Sorry, an unexpected error has occurred
Network error: Failed to fetch
Analytical Standards
Sorry, an unexpected error has occurred
Network error: Failed to fetch
REFERENCES
1.
2020. Method 537.1 Determination of Selected Per- and Polyflourinated Alkyl Substances in Drinking Water by Solid Phase Extraction and Liquid Chromatography/Tandem Mass Spectrometry (LC/MS/MS). [Internet]. Washington, DC: U.S. Environmental Protection Agency . Available from: https://cfpub.epa.gov/si/si_public_record_Report.cfm?dirEntryId=343042&Lab=NERL
2.
December 2019. Method 533 Determination of Per- and Polyfluoroalkyl Substances in Drinking Water by Isotope Dilution Anion Exchange Solid Phase Extraction and Liquid Chromatography/Tandem Mass Spectrometry. . [Internet]. United States Environmental Protection Agency, Office of Water. Available from: https://www.epa.gov/dwanalyticalmethods/method-533-determination-and-polyfluoroalkyl-substances-drinking-water-isotope
3.
August 2021. Draft Method 1633 Analysis of Per- and Polyfluoroalkyl Substances (PFAS) in Aqueous, Solid, Biosolids, and Tissue Samples by LC-MS/ MS.. [Internet]. United States Environmental Protection Agency, Office of Water. Available from: https://www.epa.gov/system/files/documents/2021-09/method_1633_draft_aug-2021.pdf
4.
December 2021. Method C-010.02 Determination of 16 Per and Polyfluoroalkyl Substances (PFAS) in Processed Food using Liquid ChromatographyTandem Mass Spectrometry (LC-MS/MS). [Internet]. United States Food and Drug Administration. Available from: https://www.fda.gov/media/131510/download
5.
October 2019. Guidelines for the Validation of Chemical Methods in Food, Feed, Cosmetics, and Veterinary Products. [Internet]. United States Food and Drug Administration. Available from: https://www.fda.gov/media/81810/download