GC Analysis of a 19-Component PCB Congener Mix on SP®-2331
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
SP-2331, 30 m x 0.25 mm I.D., 0.20 μm (24257)
oven
60 °C (1 min), 8 °C/min to 230 °C
inj. temp.
250 °C
MSD interface
220 °C
scan range
m/z = 35-500
carrier gas
helium, 1.5 mL/min constant
injection
1 μL splitless (1.0 min)
liner
4 mm I.D., single taper
sample
PCB congener mix, each analyte at 2.5 ppm in n-hexane
Description
Legal Information
SP is a registered trademark of Sigma-Aldrich Co. LLC