Merck
  • Keto-ether and glycol-ethers in the troposphere: reactivity toward OH radicals and Cl atoms, global lifetimes, and atmospheric implications.
Keto-ether and glycol-ethers in the troposphere: reactivity toward OH radicals and Cl atoms, global lifetimes, and atmospheric implications.
Environmental science and pollution research international (2017-09-25)
Javier A Barrera, Pablo R Dalmasso, Raúl A Taccone, Silvia I Lane
ABSTRACT
Rate coefficients for the gas-phase reactions of OH radicals and Cl atoms with 1-methoxy-2-propanone (1-M-2-PONE), 1-methoxy-2-propanol (1-M-2-POL), and 1-methoxy-2-butanol (1-M-2-BOL) were determined at room temperature and atmospheric pressure using a conventional relative-rate technique. The following absolute rate coefficients were derived: k
Sigma-Aldrich
1-Pentene, ≥98.5% (GC)