Merck
  • Retinoic acid receptor-related orphan receptor RORα regulates differentiation and survival of keratinocytes during hypoxia.
Retinoic acid receptor-related orphan receptor RORα regulates differentiation and survival of keratinocytes during hypoxia.
Journal of cellular physiology (2017-03-24)
Hongyu Li, Longjian Zhou, Jun Dai
ABSTRACT
Low O2 pressures present in the microenvironment of epidermis control keratinocyte differentiation and epidermal barrier function through hypoxia inducible factors (HIFs) dependent gene expression. This study focuses on investigating relations of the retinoic acid receptor-related orphan receptor alpha (RORα) to HIF-1α in keratinocytes under hypoxic conditions. The expression level of RORα is significantly elevated under hypoxia in both human and murine keratinocytes. Gene silencing of RORA attenuates hypoxia-stimulated expression of genes related to late differentiation and epidermal barrier function, and leads to an enhanced apoptotic response. While the hypoxic induction of RORα is dependent on HIF-1α, RORα is in turn critical for nuclear accumulation of HIF-1α and activation of HIF transcriptional activity. These results collectively suggest that RORα functions as an important mediator of HIF-1α activities in regulating keratinocyte differentiation/survival and epidermal barrier function during the oxygen sensing stage.
Sigma-Aldrich
Monoclonal Anti-Involucrin antibody produced in mouse, clone SY5, ascites fluid
Sigma-Aldrich
MISSION® esiRNA, targeting human RORA
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
MISSION® esiRNA, targeting human HIF1A