Merck
  • Chemical screening methods to identify ligands that promote protein stability, protein crystallization, and structure determination.
Chemical screening methods to identify ligands that promote protein stability, protein crystallization, and structure determination.
Proceedings of the National Academy of Sciences of the United States of America (2006-10-13)
Masoud Vedadi, Frank H Niesen, Abdellah Allali-Hassani, Oleg Y Fedorov, Patrick J Finerty, Gregory A Wasney, Ron Yeung, Cheryl Arrowsmith, Linda J Ball, Helena Berglund, Raymond Hui, Brian D Marsden, Pär Nordlund, Michael Sundstrom, Johan Weigelt, Aled M Edwards
ABSTRACT
The 3D structures of human therapeutic targets are enabling for drug discovery. However, their purification and crystallization remain rate determining. In individual cases, ligands have been used to increase the success rate of protein purification and crystallization, but the broad applicability of this approach is unknown. We implemented two screening platforms, based on either fluorimetry or static light scattering, to measure the increase in protein thermal stability upon binding of a ligand without the need to monitor enzyme activity. In total, 221 different proteins from humans and human parasites were screened against one or both of two sorts of small-molecule libraries. The first library comprised different salts, pH conditions, and commonly found small molecules and was applicable to all proteins. The second comprised compounds specific for protein families of particular interest (e.g., protein kinases). In 20 cases, including nine unique human protein kinases, a small molecule was identified that stabilized the proteins and promoted structure determination. The methods are cost-effective, can be implemented in any laboratory, promise to increase the success rates of purifying and crystallizing human proteins significantly, and identify new ligands for these proteins.
Sigma-Aldrich
Coenzyme B12, ≥97.0%
Sigma-Aldrich
Thiamine monophosphate chloride dihydrate
Sigma-Aldrich
Folic acid, ≥97%
Sigma-Aldrich
Pyridoxal 5′-phosphate hydrate, ≥98%
Sigma-Aldrich
Pyridoxal hydrochloride, ≥99% (HPLC)
Sigma-Aldrich
Vitamin B12, ≥98%