- Targeting YB-1 in HER-2 overexpressing breast cancer cells induces apoptosis via the mTOR/STAT3 pathway and suppresses tumor growth in mice.
Targeting YB-1 in HER-2 overexpressing breast cancer cells induces apoptosis via the mTOR/STAT3 pathway and suppresses tumor growth in mice.
The Y-box binding protein-1 (YB-1) is a transcription/translation factor that is highly expressed in primary breast tumors where it is consistently associated with poor survival. It induces human epidermal growth factor receptor (her-2) along with its dimerization partner egfr by directly binding to their promoters. In addition to promoting growth by inducing receptor tyrosine kinases, YB-1 also protects cells against apoptosis through mechanisms that have not been fully revealed. Given this, we addressed whether YB-1 might be an eventual therapeutic target for breast cancer by inhibiting it with small interfering RNAs in vitro and in vivo. Inhibiting YB-1 suppressed the growth of six of seven breast cancer cell lines that had amplified her-2 or were triple negative. Importantly, targeting YB-1 induced apoptosis in BT474-m1 and Au565 breast cancer cells known to have her-2 amplifications. The potential role of signal transducers and activators of transcription 3 (STAT3) was pursued to address the underlying mechanism for YB-1-mediated survival. Inhibition of YB-1 decreased P-STAT3(S727) but not P-STAT3(Y705) or total STAT3. This was accompanied by decreased P-ERK1/2(T202/Y204), P-mTOR(S2448), and total mammalian target of rapamycin mTOR. Furthering the role of STAT3 in these cells, we show that knocking it down recapitulated the induction of apoptosis. Alternatively, constitutively active P-STAT3 rescued YB-1-induced apoptosis. Finally, targeting YB-1 with 2 different siRNAs remarkably suppressed tumor cell growth in soft agar by >90% and delayed tumorigenesis in nude mice. We conclude that HER-2 overexpressing as well as triple-negative breast cancer cells are YB-1 dependent, suggesting it may be a good therapeutic target for these exceptionally aggressive tumors.