Merck
Kinetics of the Reactions of NO
The journal of physical chemistry. A (2017-05-13)
Li Zhou, A R Ravishankara, Steven S Brown, Mahmoud Idir, Kyle J Zarzana, Véronique Daële, Abdelwahid Mellouki
ABSTRACT
Two different experimental methods (relative rate and absolute rate methods) were used to measure the rate coefficients for the reactions of NO
Sigma-Aldrich
Propylene, ≥99%
Sigma-Aldrich
Isobutyl methacrylate, 97%
Sigma-Aldrich
Propionaldehyde, natural, ≥98%, FG
Sigma-Aldrich
Isobornyl methacrylate, technical grade