Merck
  • Synthesis of Heterotelechelic Polymers for Conjugation of Two Different Proteins.
Synthesis of Heterotelechelic Polymers for Conjugation of Two Different Proteins.
Macromolecules (2009-01-01)
Karina L Heredia, Gregory N Grover, Lei Tao, Heather D Maynard
ABSTRACT
In this report we describe a straightforward approach to synthesize polymers with end-groups that bind site-specifically to two different proteins. Telechelic biotin, maleimide poly(N-isopropylacrylamide) (pNIPAAm) was synthesized for the formation of streptavidin (SAv)-bovine serum albumin (BSA) polymer conjugates. Reversible addition-fragmentation chain transfer (RAFT) polymerization of NIPAAm was conducted in the presence of biotinylated chain transfer agents (CTAs) with either ester or amide linkages, and the resultant α-biotinylated pNIPAAm were formed with low polydispersity indices (PDI ≤ 1.09). UV-Vis analysis of the trithiocarbonate chain-ends indicated 88% or greater retention of the group. A maleimide was introduced to the ω chain-end via a radical cross-coupling reaction with a functionalized azo-initiator. The polymer structures were characterized by 1H NMR spectroscopy and gel permeation chromatography (GPC). The resultant biotin-maleimide heterotelechelic polymer was used to form a SAv-BSA heterodimer conjugate. Bioconjugate formation was confirmed by gel electrophoresis.
Sigma-Aldrich
11-Maleimidoundecanoic acid, 95% (GC)
Sigma-Aldrich
Poly(ethylene glycol) methyl ether maleimide, average Mn 2,000